accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 155206-00-1

IUPAC Name: (5Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3S)-3-hydroxy-5-phenylpent-1-en-1-yl]cyclopentyl]-N-ethylhept-5-enamide
Synonyms: bimatoprost lumigan bimataprost bimatoprostum
Molecular Weight (g/mol): 415.57
Molecular Formula: C25H37NO4
InChi Key: AQOKCDNYWBIDND-FTOWTWDKSA-N
SMILES: CCNC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\[C@@H](O)CCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results