accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15522-59-5

IUPAC Name: tetrabutylazanium; tetraphenylboranuide
Synonyms: tetrabutylammonium ion tetraphenylborate
Molecular Weight (g/mol): 561.71
Molecular Formula: C40H56BN
InChi Key: ZHCCBGAUZWZGQV-UHFFFAOYSA-N
SMILES: CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results