accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15525-64-1

IUPAC Name: silver(1+) 2,4-dioxopentan-3-ide
Synonyms: silver(1+) acetylacetonate
Molecular Weight (g/mol): 206.98
Molecular Formula: C5H7AgO2
InChi Key: UEGHNEVWUGTMES-UHFFFAOYSA-N
SMILES: [Ag+].CC(=O)[CH-]C(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results