accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CFF

CAS RN 155266-68-5

IUPAC Name: 4-(but-3-en-1-yl)-4'-(3,4-difluorophenyl)-1,1'-bi(cyclohexane)
Synonyms: 4-(but-3-en-1-yl)-4'-(3,4-difluorophenyl)-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 332.48
Molecular Formula: C22H30F2
InChi Key: UMULWEQZNFZORL-UHFFFAOYSA-N
SMILES: FC1=C(F)C=C(C=C1)C1CCC(CC1)C1CCC(CCC=C)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results