accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 155306-71-1

IUPAC Name: 5-(1,3-benzothiazol-2-yl)-10,10,16,16-tetramethyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Synonyms: coumarin 545T
Molecular Weight (g/mol): 430.57
Molecular Formula: C26H26N2O2S
InChi Key: MSDMPJCOOXURQD-UHFFFAOYSA-N
SMILES: CC1(C)CCN2CCC(C)(C)C3=C4OC(=O)C(=CC4=CC1=C23)C1=NC2=CC=CC=C2S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results