accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CFPFFFFF

CAS RN 155371-19-0

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-ethyl-3-methylimidazolium hexafluorophosphate
Molecular Weight (g/mol): 256.13
Molecular Formula: C6H11F6N2P
InChi Key: DPDAKOVGQUGTHH-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCN1C=C[N+](C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)