accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 155395-45-2

IUPAC Name: 2-(furan-2-yl)benzonitrile
Synonyms: 2-(furan-2-yl)benzonitrile
Molecular Weight (g/mol): 169.18
Molecular Formula: C11H7NO
InChi Key: XLZNEBADSPRURH-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=C1C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results