accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15546-75-5

IUPAC Name: 5,10-dimethyl-5,10-dihydrophenazine
Synonyms: 5,10-dimethylphenazine
Molecular Weight (g/mol): 210.28
Molecular Formula: C14H14N2
InChi Key: GVTGSIMRZRYNEI-UHFFFAOYSA-N
SMILES: CN1C2=CC=CC=C2N(C)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results