CAS RN 1556-34-9
IUPAC Name:
9-(bromomethyl)acridine
Synonyms:
9-(bromomethyl)acridine
Molecular Weight (g/mol):
272.15
Molecular Formula:
C14H10BrN
InChi Key:
MZFYKBHQWLWIBI-UHFFFAOYSA-N
SMILES:
BrCC1=C2C=CC=CC2=NC2=CC=CC=C12
1
–
1
of
1
results
1
9-(Bromomethyl)acridine 98.0+%, TCI America™
CAS: 1556-34-9 Molecular Formula: C14H10BrN Molecular Weight (g/mol): 272.15 MDL Number: MFCD01321146 InChI Key: MZFYKBHQWLWIBI-UHFFFAOYSA-N PubChem CID: 150930 IUPAC Name: 9-(bromomethyl)acridine SMILES: BrCC1=C2C=CC=CC2=NC2=CC=CC=C12