accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 155601-71-1

IUPAC Name: tert-butyl 6-fluoro-2H-pyrazolo[3,4-b]pyridine-3-carboxylate
Synonyms: tert-butyl 6-fluoro-2H-pyrazolo[3,4-b]pyridine-3-carboxylate
Molecular Weight (g/mol): 237.23
Molecular Formula: C11H12FN3O2
InChi Key: INGAFDNQIOIZHC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=C2C=CC(F)=NC2=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results