CAS RN 155633-54-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Drometrizole trisiloxane, ≥98% (HPLC), MilliporeSigma™ Supelco™
Drometrizole trisiloxane (DRT) belongs to the lipophilic benzotriazole derivative class of compounds commonly utilized in sunscreen cosmetics.