accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15570-45-3

IUPAC Name: (2,4,5-triphenylcyclopenta-1,4-dien-1-yl)benzene
Synonyms: (2,4,5-triphenylcyclopenta-1,4-dien-1-yl)benzene
Molecular Weight (g/mol): 370.5
Molecular Formula: C29H22
InChi Key: JCXLYAWYOTYWKM-UHFFFAOYSA-N
SMILES: C1C(=C(C(=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)