CAS RN 155751-72-7
IUPAC Name:
potassium 2-hydroxy-3-(5-hydroxy-8-{4-[8'-hydroxy-6'-(1-hydroxy-3-{3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl}butyl)-7'-methylidene-hexahydro-3'H-spiro[oxolane-2,2'-pyrano[3,2-b]pyran]-5-yl]but-3-en-2-yl}-10-methyl-1,7-dioxaspiro[5.5]undec-10-en-2-yl)-2-methylpropanoate
Synonyms:
potassium 2-hydroxy-3-(5-hydroxy-8-{4-[8'-hydroxy-6'-(1-hydroxy-3-{3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl}butyl)-7'-methylidene-hexahydrospiro[oxolane-2,2'-pyrano[3,2-b]pyran]-5-yl]but-3-en-2-yl}-10-methyl-1,7-dioxaspiro[5.5]undec-10-en-2-yl)-2-methylpropanoate
Molecular Weight (g/mol):
843.11
Molecular Formula:
C44H67KO13
InChi Key:
UXRQUXBFVICHQJ-UHFFFAOYNA-M
SMILES:
[K+].CC(CC(O)C1OC2CCC3(CCC(O3)C=CC(C)C3CC(C)=CC4(OC(CC(C)(O)C([O-])=O)CCC4O)O3)OC2C(O)C1=C)C1OC2(CCCCO2)CCC1C
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Okadaic Acid, Potassium Salt
Water-soluble analog of Okadaic Acid. Inhibits protein phosphatases 1 and 2A.