accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 155773-67-4

IUPAC Name: copper(2+) 5,12,18,25,31,38,44,51-octabutoxy-2,15,28,41,53,54,55,56-octaazatridecacyclo[40.10.1.1³,¹⁴.1¹⁶,²⁷.1²⁹,⁴⁰.0⁴,¹³.0⁶,¹¹.0¹⁷,²⁶.0¹⁹,²⁴.0³⁰,³⁹.0³²,³⁷.0⁴³,⁵².0⁴⁵,⁵⁰]hexapentaconta-1(52),2,4,6,8,10,12,14(56),15,17,19,21,23,25,27,29(54),30,32,34,36,38,40,42,44,46,48,50-heptacosaene-53,55-diide
Synonyms: copper(2+) 5,12,18,25,31,38,44,51-octabutoxy-2,15,28,41,53,54,55,56-octaazatridecacyclo[40.10.1.1³,¹⁴.1¹⁶,²⁷.1²⁹,⁴⁰.0⁴,¹³.0⁶,¹¹.0¹⁷,²⁶.0¹⁹,²⁴.0³⁰,³⁹.0³²,³⁷.0⁴³,⁵².0⁴⁵,⁵⁰]hexapentaconta-1(52),2,4,6,8,10,12,14(56),15,17,19,21,23,25,27,29(54),30,32,34,36,38,40,42,44,46,48,50-heptacosaene-53,55-diide
Molecular Weight (g/mol): 1353.18
Molecular Formula: C80H88CuN8O8
InChi Key: JJIWQZDKAOMTKU-UHFFFAOYSA-N
SMILES: [Cu++].CCCCOC1=C2C3=NC4=NC(=NC5=C6C(=C([N-]5)N=C5N=C(N=C([N-]3)C2=C(OCCCC)C2=CC=CC=C12)C1=C(OCCCC)C2=CC=CC=C2C(OCCCC)=C51)C(OCCCC)=C1C=CC=CC1=C6OCCCC)C1=C(OCCCC)C2=CC=CC=C2C(OCCCC)=C41
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results