accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15578-26-4

IUPAC Name: bis(λ²-tin(2+)) (phosphonatooxy)phosphonate
Synonyms: stannous pyrophosphate
Molecular Weight (g/mol): 411.36
Molecular Formula: O7P2Sn2
InChi Key: GEZAUFNYMZVOFV-UHFFFAOYSA-J
SMILES: [Sn++].[Sn++].[O-]P([O-])(=O)OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)