accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 155836-50-3

IUPAC Name: 4-{[2,5-dimethyl-4-(prop-2-en-1-yl)piperazin-1-yl](3-hydroxyphenyl)methyl}-N,N-diethylbenzamide
Synonyms: 4-{[2,5-dimethyl-4-(prop-2-en-1-yl)piperazin-1-yl](3-hydroxyphenyl)methyl}-N,N-diethylbenzamide
Molecular Weight (g/mol): 435.61
Molecular Formula: C27H37N3O2
InChi Key: LBLDMHBSVIVJPM-UHFFFAOYNA-N
SMILES: CCN(CC)C(=O)C1=CC=C(C=C1)C(N1CC(C)N(CC=C)CC1C)C1=CC(O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results