accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHFF

CAS RN 155997-19-6

IUPAC Name: N-(2,6-difluorophenyl)-4-methoxybenzamide
Synonyms: N-(2,6-difluorophenyl)-4-methoxybenzamide
Molecular Weight (g/mol): 263.24
Molecular Formula: C14H11F2NO2
InChi Key: NDUMEFRGYXBRDG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)NC1=C(F)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results