accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOH

CAS RN 156-39-8

IUPAC Name: 3-(4-hydroxyphenyl)-2-oxopropanoic acid
Synonyms: 4-hydroxyphenylpyruvic acid (p-hydroxyphenyl)pyruvic acid (4-hydroxyphenyl)pyruvic acid
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: KKADPXVIOXHVKN-UHFFFAOYSA-N
SMILES: OC(=O)C(=O)CC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)