accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl(E)

CAS RN 156-60-5

IUPAC Name: (1E)-1,2-dichloroethene
Synonyms: trans-1,2-dichloroethylene dioform 1,2-dichloroethene (substance) 1,2-trans-dichloroethylene dichloro-1,2-ethylene 1,2-dichloroethene (trans) rcra waste number U079 1,2-dichlor-aethen
Molecular Weight (g/mol): 96.94
Molecular Formula: C2H2Cl2
InChi Key: KFUSEUYYWQURPO-OWOJBTEDSA-N
SMILES: Cl\C=C\Cl
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
  • (2)
Quantity 
  • (2)
  • (4)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)