accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH2P

CAS RN 1560-54-9

IUPAC Name: triphenyl(prop-2-en-1-yl)phosphanium bromide
Synonyms: triphenyl(prop-2-en-1-yl)phosphanium bromide
Molecular Weight (g/mol): 383.27
Molecular Formula: C21H20BrP
InChi Key: FWYKRJUVEOBFGH-UHFFFAOYSA-M
SMILES: [Br-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)