CAS RN 1561-11-1
IUPAC Name:
4-methylhexanoic acid
Synonyms:
4-methylhexanoic acid
Molecular Weight (g/mol):
130.19
Molecular Formula:
C7H14O2
InChi Key:
DIVCBWJKVSFZKJ-UHFFFAOYNA-N
SMILES:
CCC(C)CCC(O)=O
1
–
1
of
1
results
1
Sigma Aldrich ™-(-)-4-Phenyl-2-Butanol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.