accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNHN(Z)

CAS RN 1562-95-4

IUPAC Name: 2-{[(2Z)-1,2-dihydropyridin-2-ylidene](nitroso)methyl}pyridine
Synonyms: 2-[nitroso((2Z)-1H-pyridin-2-ylidene)methyl]pyridine
Molecular Weight (g/mol): 199.21
Molecular Formula: C11H9N3O
InChi Key: RZHHBDXWQUIRSQ-LUAWRHEFSA-N
SMILES: O=N\C(=C1/NC=CC=C1)C1=CC=CC=N1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results