accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 156270-06-3

IUPAC Name: 1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
Synonyms: 1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: KYBIRFFGAIFLPM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNC2=NC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results