accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1563-01-5

IUPAC Name: 3-{2-[(1E)-4-(diethylamino)-6-oxocyclohexa-2,4-dien-1-ylidene]hydrazin-1-yl}-4-hydroxybenzene-1-sulfonic acid
Synonyms: 3-{2-[(1E)-4-(diethylamino)-6-oxocyclohexa-2,4-dien-1-ylidene]hydrazin-1-yl}-4-hydroxybenzenesulfonic acid
Molecular Weight (g/mol): 365.4
Molecular Formula: C16H19N3O5S
InChi Key: DGIJTEMQAXADQJ-GHRIWEEISA-N
SMILES: CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC(=CC=C1O)S(O)(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results