accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1563-38-8

IUPAC Name: 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-ol
Synonyms: carbofuran phenol carbofuran 7-phenol carbofuranphenol
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: WJGPNUBJBMCRQH-UHFFFAOYSA-N
SMILES: CC1(C)CC2=CC=CC(O)=C2O1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results