accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaNaOHHOOHHOOHHOOOOOOO

CAS RN 15630-89-4

IUPAC Name: tetrasodium tris(peroxol) dicarbonate
Synonyms: sodium percarbonate
Molecular Weight (g/mol): 314.02
Molecular Formula: C2H6Na4O12
InChi Key: MSLRPWGRFCKNIZ-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].OO.OO.OO.[O-]C([O-])=O.[O-]C([O-])=O
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
Grade 
  • (6)
Physical Form 
  • (2)
  • (4)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (4)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
  • (6)
  • (2)
  • (4)
  • (2)