accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 156492-30-7

IUPAC Name: 4,7-dibromo-1,10-phenanthroline
Synonyms: 4,7-dibromo-1,10-phenanthroline
Molecular Weight (g/mol): 338
Molecular Formula: C12H6Br2N2
InChi Key: AKZAIDYHEKUXBU-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC3=C(Br)C=CN=C3C2=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results