accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSNH2OO

CAS RN 1565-17-9

IUPAC Name: 4-acetylbenzene-1-sulfonamide
Synonyms: P-acetylbenzenesulfonamide
Molecular Weight (g/mol): 199.22
Molecular Formula: C8H9NO3S
InChi Key: CSATVXJBGFVJES-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results