accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15656-28-7

IUPAC Name: bis(pyridin-1-ium-1-yl)iodanuide; tetrafluoroboranuide
Synonyms: bis(pyridin-1-ium-1-yl)iodanuide tetrafluoroborate
Molecular Weight (g/mol): 371.91
Molecular Formula: C10H10BF4IN2
InChi Key: BMDSRCBKJZCUBH-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.[I-]([N+]1=CC=CC=C1)[N+]1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (4)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (5)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)