accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 156567-57-6

IUPAC Name: bromozincylium; propan-1-ide
Synonyms: bromozincylium; propan-1-ide
Molecular Weight (g/mol): 188.37
Molecular Formula: C3H7BrZn
InChi Key: BXIZKCIGQKZYGR-UHFFFAOYSA-M
SMILES: [Zn+]Br.CC[CH2-]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results