accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHO

CAS RN 15658-11-4

IUPAC Name: 4-(hydroxydiphenylmethyl)phenol
Synonyms: 4-(hydroxydiphenylmethyl)phenol
Molecular Weight (g/mol): 276.34
Molecular Formula: C19H16O2
InChi Key: QEIRHBKKYVWAOM-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results