accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1566-42-3

IUPAC Name: (4-methoxyphenyl)urea
Synonyms: urea, (4-methoxyphenyl)-
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: PGUKYDVWVXRPKK-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(N)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results