accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NNHNOONOO

CAS RN 1567-89-1

IUPAC Name: 1-(2,4-dinitrophenyl)-2-(propan-2-ylidene)hydrazine
Synonyms: 1-(2,4-dinitrophenyl)-2-(propan-2-ylidene)hydrazine
Molecular Weight (g/mol): 238.2
Molecular Formula: C9H10N4O4
InChi Key: YGIXYAIGWMAGIB-UHFFFAOYSA-N
SMILES: CC(C)=NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Grade 
  • (3)
Industry Type 
  • (1)
  • (2)
Solvent or Matrix 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)