accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNCH3OHO

CAS RN 15674-67-6

IUPAC Name: hydrogen 3-(diethylamino)propanoic acid chloride
Synonyms: hydrogen 3-(diethylamino)propanoic acid chloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: QQVHFNAGPZTCBE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results