accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrBrF

CAS RN 156772-60-0

IUPAC Name: 2,5-dibromo-3-fluoropyridine
Synonyms: 2,5-dibromo-3-fluoropyridine
Molecular Weight (g/mol): 254.88
Molecular Formula: C5H2Br2FN
InChi Key: QEENLQKOTDHQEW-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CN=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results