accessibility menu, dialog, popup

UserName

Viewing profile as user:
PPClClPtH3CH3CH3CH3CCH3H3C

CAS RN 15692-07-6

IUPAC Name: platinum(2+) bis(triethylphosphane) dichloride
Synonyms: platinum(2+) bis(triethylphosphine) dichloride
Molecular Weight (g/mol): 502.3
Molecular Formula: C12H30Cl2P2Pt
InChi Key: WPWLTKRUFHHDLP-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pt++].CCP(CC)CC.CCP(CC)CC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)