accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COON

CAS RN 1570-45-2

IUPAC Name: ethyl pyridine-4-carboxylate
Synonyms: isonicotinic acid, ethyl ester
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: MCRPKBUFXAKDKI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=NC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)