accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNClFFF

CAS RN 157021-61-9

IUPAC Name: 2,6-dichloro-4-(trifluoromethyl)benzonitrile
Synonyms: 2,6-dichloro-4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 240.01
Molecular Formula: C8H2Cl2F3N
InChi Key: NCXSSFQXQAOREM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(C#N)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results