accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNH3CH3CCH3CH3CH3CH3H3COVHClHCl

CAS RN 15709-03-2

IUPAC Name: 5,10,15,20-tetraethyl-4,9,14,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11(23),12,14,16,18-undecaene-21,22-diide oxovanadium dihydrochloride
Synonyms: 5,10,15,20-tetraethyl-4,9,14,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11(23),12,14,16,18-undecaene-21,22-diide vanadium monoxide dihydrochloride
Molecular Weight (g/mol): 616.53
Molecular Formula: C32H38Cl2N4OV
InChi Key: LLVNESQRTPCYSP-UHFFFAOYSA-N
SMILES: Cl.Cl.O=[V].CCC1=C(C)C2=CC3=NC(=CC4=NC(=CC5=C(CC)C(C)=C([N-]5)C=C1[N-]2)C(C)=C4CC)C(C)=C3CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results