accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOHO

CAS RN 1571-30-8

IUPAC Name: 8-hydroxyquinoline-2-carboxylate
Synonyms: 8-hydroxyquinoline-2-carboxylate
Molecular Weight (g/mol): 188.16
Molecular Formula: C10H6NO3
InChi Key: UHBIKXOBLZWFKM-UHFFFAOYSA-M
SMILES: OC1=C2N=C(C=CC2=CC=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)