accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1571-33-1

IUPAC Name: phenylphosphonic acid
Synonyms: phenylphosphonic acid
Molecular Weight (g/mol): 158.09
Molecular Formula: C6H7O3P
InChi Key: QLZHNIAADXEJJP-UHFFFAOYSA-N
SMILES: OP(O)(=O)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (4)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (3)
  • (6)
  • (4)
  • (1)
  • (1)
  • (3)
  • (6)