accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15710-66-4

IUPAC Name: manganese(2+) dinitrate
Synonyms: manganese(II) nitrate
Molecular Weight (g/mol): 178.95
Molecular Formula: MnN2O6
InChi Key: MIVBAHRSNUNMPP-UHFFFAOYSA-N
SMILES: [Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)