accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NNCH3CH3CCH3

CAS RN 157197-53-0

IUPAC Name: 1,3-di-tert-butyl-1H-imidazol-3-ium-2-ide
Synonyms: 1,3-di-tert-butylimidazol-1-ium-2-ide
Molecular Weight (g/mol): 180.3
Molecular Formula: C11H20N2
InChi Key: FENRCIKTFREPGS-UHFFFAOYSA-N
SMILES: CC(C)(C)N1C=C[N+](=[C-]1)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results