accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1572-10-7

IUPAC Name: 5-phenyl-1H-pyrazol-3-amine
Synonyms: 5-phenyl-1H-pyrazol-3-amine
Molecular Weight (g/mol): 159.19
Molecular Formula: C9H9N3
InChi Key: PWSZRRFDVPMZGM-UHFFFAOYSA-N
SMILES: NC1=NNC(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)