accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH3CO

CAS RN 1573-17-7

IUPAC Name: 4-(acetyloxy)but-2-yn-1-yl acetate
Synonyms: 4-(acetyloxy)but-2-yn-1-yl acetate
Molecular Weight (g/mol): 170.16
Molecular Formula: C8H10O4
InChi Key: TVIMIQVBDDNCCR-UHFFFAOYSA-N
SMILES: CC(=O)OCC#CCOC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results