accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15743-20-1

IUPAC Name: aluminium(3+) tris(prop-2-enoate)
Synonyms: aluminium(3+) triacrylate
Molecular Weight (g/mol): 240.15
Molecular Formula: C9H9AlO6
InChi Key: LIQDVINWFSWENU-UHFFFAOYSA-K
SMILES: [Al+3].[O-]C(=O)C=C.[O-]C(=O)C=C.[O-]C(=O)C=C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results