accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15746-57-3

IUPAC Name: λ²-ruthenium(2+) bis(2,2'-bipyridine) dihydrate dichloride
Synonyms: λ²-ruthenium(2+) bis(bipyridyl) dihydrate dichloride
Molecular Weight (g/mol): 520.38
Molecular Formula: C20H20Cl2N4O2Ru
InChi Key: NHKTUSUPCAKVHT-UHFFFAOYSA-L
SMILES: O.O.[Cl-].[Cl-].[Ru++].C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(N=C1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results