accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15750-47-7

IUPAC Name: dicerium(3+) trioxalate
Synonyms: cerium oxalate|cerium oxalate anhydrous
Molecular Weight (g/mol): 544.29
Molecular Formula: C6Ce2O12
InChi Key: ZMZNLKYXLARXFY-UHFFFAOYSA-H
SMILES: [Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)