accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 157599-02-5

IUPAC Name: 2-[4,7,10-tris(carbamoylmethyl)-1,4,7,10-tetraazacyclododecan-1-yl]acetamide
Synonyms: 2-[4,7,10-tris(carbamoylmethyl)-1,4,7,10-tetraazacyclododecan-1-yl]acetamide
Molecular Weight (g/mol): 400.48
Molecular Formula: C16H32N8O4
InChi Key: FQIHLPGWBOBPSG-UHFFFAOYSA-N
SMILES: NC(=O)CN1CCN(CC(N)=O)CCN(CC(N)=O)CCN(CC(N)=O)CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)