accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH(Z)

CAS RN 1576-95-0

IUPAC Name: (2Z)-pent-2-en-1-ol
Synonyms: cis-2-pentenol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: BTSIZIIPFNVMHF-ARJAWSKDSA-N
SMILES: CC\C=C/CO
Boiling Point 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)